4-Fluorobenzaldehyde, 98+%
4-Fluorobenzaldehyde, 98+%
4-Fluorobenzaldehyde, 98+%
Thermo Scientific Chemicals

4-Fluorobenzaldehyde, 98+%

CAS: 459-57-4 | C7H5FO | 124.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 156150010
Price (USD)
787.65
Special offer
Online exclusive
Ends: 28-Dec-2026
926.00
Save 138.35
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
787.65
Special offer
Online exclusive
Ends: 28-Dec-2026
926.00
Save 138.35
Each
Chemical Identifiers
CAS459-57-4
IUPAC Name4-fluorobenzaldehyde
Molecular FormulaC7H5FO
InChI KeyUOQXIWFBQSVDPP-UHFFFAOYSA-N
SMILESFC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
GC>=98.0 %
Infrared spectrumConforms
Refractive index1.5195 to 1.5215 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only