Supercoiled DNA Ladder (for sizing supercoiled DNA) - FAQs
Supercoiled DNA Ladder (for sizing supercoiled DNA) - FAQs
View additional product information for Supercoiled DNA Ladder (for sizing supercoiled DNA) - FAQs (15622012)
1 product FAQs found
Do you offer an alternative for the discontinued Supercoiled DNA Ladder (for sizing supercoiled DNA) (Cat. No. 15622-012)?
We no longer offer supercoiled DNA ladders. We recommend linearizing DNA first.
Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.