0.5-10 Kb RNA Ladder - FAQs
0.5-10 Kb RNA Ladder - FAQs
View additional product information for 0.5-10 Kb RNA Ladder - FAQs (15623200)
2 product FAQs found
Why are my RNA bands not sharp?
(1) RNA was not completely denatured. Electrophorese RNA under denaturing conditions. Use urea, formamide or formaldehyde gels, or glyoxal-treated RNA.
(2) For glyoxal-treated RNA, a buffer gradient formed during electrophoresis. Recirculate buffer during electrophoresis to prevent gradient formation.
Why are the RNA bands disappearing when looking and photographing a gel on a UV box?
RNA was exposed to UV light for extended periods of time. Minimize exposure to UV light. Stain and destain gels in the dark and photograph the gel immediately.