2-Hexanol, 99%
2-Hexanol, 99%
2-Hexanol, 99%
Thermo Scientific Chemicals

2-Hexanol, 99%

CAS: 626-93-7 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 156290100
Price (USD)
36.65
Online exclusive
41.10
Save 4.45
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
36.65
Online exclusive
41.10
Save 4.45
Each
Chemical Identifiers
CAS626-93-7
IUPAC Namehexan-2-ol
Molecular FormulaC6H14O
InChI KeyQNVRIHYSUZMSGM-UHFFFAOYNA-N
SMILESCCCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=98.5 %
Refractive index1.4125 to 1.4155 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only