HEPES (1M) - FAQs
HEPES (1M) - FAQs
View additional product information for HEPES (1M) - FAQs (15630056)
2 product FAQs found
Is HEPES (1 M) in distilled water or in saline solution?
1M HEPES is in water with a pH range of 7.2 - 7.5.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the endotoxin specification for HEPES (1 M) (Cat. Nos. 15630080, 15630106, and 15630130)?
Sorry, we do not test endotoxin levels for this product.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.