HEPES (1 M)
Gibco™

HEPES (1 M)

Gibco HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. It is membrane impermeable, relatively stable, and has limited interference in key processes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1563013020 x 100 mL
1563010620 mL
15630080100 mL
Catalog number 15630130
Price (USD)
1,458.65
Online exclusive
1,586.00
Save 127.35
Each
Quantity:
20 x 100 mL
Price (USD)
1,458.65
Online exclusive
1,586.00
Save 127.35
Each

Gibco HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. The addition of 10–25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator.

Gibco HEPES (N-2-hydroxyethylpiperazine-N-2-ethane sulfonic acid) is a good buffering choice for many cell culture systems because it:

  • Is membrane impermeable
  • Has limited effect on biochemical reactions
  • Is chemically and enzymatically stable
  • Has a very low visible and UV light absorbance

cGMP manufacturing and quality system

For supply chain continuity, we manufacture this product at two separate facilities located in Grand Island, NY, and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS7365-45-9
Chemical Name or MaterialZwitterionic Organic Chemical Buffer
Recommended StorageStorage conditions: 2°C to 8°C, Shipping conditions: Room temperature, Shelf life: 24 months from date of manufacture
SterilitySterile
Sterilization MethodSterile-filtered
Concentration1 M
For Use With (Application)Chromatin Biology
GradeBiochemical
Molecular Weight (g/mol)238.3 g/mol
Physical FormLiquid
Quantity20 x 100 mL
Solution TypeBuffer
pH7 to 7.5
Unit SizeEach

Frequently asked questions (FAQs)

My Gibco HEPES (1M) (Cat. No. 15630080) was accidentally frozen. Can I still use it?

This product should be stored at 2-8 degrees C and should not be frozen. The biggest issue with accidentally freezing this product is its solubility. If this product was accidentally frozen, we recommend placing it in a 2-8 degree environment and allowing it to slowly thaw overnight. If the product is fully thawed and shows no signs of precipitation, then it should still be usable, but we cannot guarantee effectiveness.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is HEPES (1 M) in distilled water or in saline solution?

1M HEPES is in water with a pH range of 7.2 - 7.5.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the endotoxin specification for HEPES (1 M) (Cat. Nos. 15630080, 15630106, and 15630130)?

Sorry, we do not test endotoxin levels for this product.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (2)

Citations & References
Abstract
Preparation and characterization of toxic Abeta aggregates for structural and functional studies in Alzheimer's disease research.
Authors:Jan A, Hartley DM, Lashuel HA,
Journal:Nat Protoc
PubMed ID:20539293
'The amyloid cascade hypothesis, supported by strong evidence from genetics, pathology and studies using animal models, implicates amyloid-beta (Abeta) oligomerization and fibrillogenesis as central causative events in the pathogenesis of Alzheimer''s disease (AD). Today, significant efforts in academia, biotechnology and the pharmaceutical industry are devoted to identifying the mechanisms by ... More
Investigating conversion of mechanical force into biochemical signaling in three-dimensional chondrocyte cultures.
Authors:Bougault C, Paumier A, Aubert-Foucher E, Mallein-Gerin F,
Journal:Nat Protoc
PubMed ID:19478808
The culture of chondrocytes embedded within agarose hydrogels maintains chondrocytic phenotype over extended periods and allows analysis of the chondrocyte response to mechanical forces. The mechanisms involved in the transduction of a mechanical stimulus to a physiological process are not completely deciphered. We present protocols to prepare and characterize constructs ... More