UltraPure™ Salmon Sperm DNA Solution is a ready-to-use, sheared DNA solution that is used directly in the preparation of prehybridizationRead more
Have Questions?
Catalog Number
Quantity
15632011
5 x 1 mL
Catalog number 15632011
Price (USD)
230.65
Online exclusive
254.00
Save 23.35
Each
Quantity:
5 x 1 mL
Request bulk or custom format
Price (USD)
230.65
Online exclusive
254.00
Save 23.35
Each
UltraPure™ Salmon Sperm DNA Solution is a ready-to-use, sheared DNA solution that is used directly in the preparation of prehybridization and hybridization solutions. This DNA solution is prepared from highly pure, phenol/chloroform-extracted DNA and DNase-free, RNase-free (DEPC-treated), distilled, deionized water. The DNA is sheared to an average size of ≤2,000 bp. The concentration is adjusted to 10 mg/mL.
Applications Such solutions are used to block the nonspecific attachment of probe to the surface of a membrane.
Performance and quality testing Concentration, size verification, viscosity, and ultraviolet absorbance (A260/A280) are analytically tested.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Chromatin Biology, Nucleic Acid Gel Electrophoresis, Blotting
Key FunctionsBlock Non-Specific Hybridization
Product LineUltraPure
Product TypeSalmon Sperm DNA Solution
Quantity5 x 1 mL
Reagent TypeSalmon Sperm DNA
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
5 tubes Salmon Sperm DNA solution (1 ml each, 10 mg/ml)
UltraPure™ Salmon Sperm DNA is stable for 1 year at -20°C.
Frequently asked questions (FAQs)
What species of salmon is Ultrapure Salmon Sperm DNA derived from?
The species of salmon is Oncerhyncus keta.
Is Ultrapure Salmon Sperm DNA double stranded? How can I make it single stranded?
Ultrapure Salmon Sperm DNA is double stranded. It can be made single stranded by heating to 95 degrees C and then cooling rapidly on ice. Cooling is usually done with the DNA diluted in a hybridization solution which keeps it single stranded because of the large dilution volume.
What concentration should the UltraPure Salmon Sperm DNA Solution (Cat. No. 15632011) be used at?
Carrier DNA is typically used at a concentration of 100 µg/mL in both the prehybridization and hybridization solutions.
Citations & References (8)
Citations & References
Abstract
The human organic cation transporter (hOCT2) recognizes the degree of substrate ionization.
Authors: Barendt Wendy M; Wright Stephen H;
Journal:J Biol Chem
PubMed ID:11953440
'The organic cation transporter, OCT2, plays a role in renal secretion of a broad array of weak bases. To determine whether the degree of ionization of these compounds plays a role in their interaction with OCT2, we examined the influence of external pH values on the activity of the human ... More
The role of polymer nanolayer architecture on the separation performance of anion-exchange membrane adsorbers: part II. DNA and virus separations.
Authors:Bhut BV, Weaver J, Carter AR, Wickramasinghe SR, Husson SM,
Journal:Biotechnol Bioeng
PubMed ID:21618476
'The surface-initiated polymerization protocol developed in part I was used to prepare strong anion-exchange membranes with variable polymer chain graft densities and degrees of polymerization for DNA and virus particle separations. A focus of part II was to evaluate the role of polymer nanolayer architecture on DNA and virus binding. ... More
Spectrophotometric analysis of nucleic acids: oxygenation-dependent hyperchromism of DNA.
Authors:Doshi R, Day PJ, Carampin P, Blanch E, Stratford IJ, Tirelli N,
Journal:Anal Bioanal Chem
PubMed ID:20169336
'The absorbance at 260 nm (A(260)) is ubiquitously used for nucleic acid quantification. We show that following oxygenation, DNA solutions experience alterations in both spectral properties (hyperchromism in the UV region, lambda(max) 260 nm) and DNA conformation. The spectral changes caused by oxygen-DNA complexation are stable for at least several ... More
A quantitative real-time polymerase chain reaction assay for the seagrass pathogen Labyrinthula zosterae.
'The protist Labyrinthula zosterae (Phylum Bigyra, sensu Tsui et al. 2009) has been identified as a causative agent of wasting disease in eelgrass (Zostera marina), of which the most intense outbreak led to the destruction of 90% of eelgrass beds in eastern North America and western Europe in the 1930s. ... More
Noncoding DNA in lipofection of HeLa cells-a few insights.
Authors:Symens N, Rejman J, Lucas B, Demeester J, De Smedt SC, Remaut K,
Journal:Mol Pharm
PubMed ID:23421924
'In cationic carrier-mediated gene delivery, the disproportional relationship between the quantity of delivered DNA and the amount of encoded protein produced is a well-known phenomenon. The numerous intracellular barriers which need to be overcome by pDNA to reach the nucleoplasm play a major role in it. In contrast to what ... More