2-Hydroxyethyl methacrylate, 97%, stabilized
2-Hydroxyethyl methacrylate, 97%, stabilized
2-Hydroxyethyl methacrylate, 97%, stabilized
Thermo Scientific Chemicals

2-Hydroxyethyl methacrylate, 97%, stabilized

CAS: 868-77-9 | C6H10O3 | 130.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 156330010
Price (USD)
91.50
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
91.50
Each
Chemical Identifiers
CAS868-77-9
IUPAC Name2-hydroxyethyl 2-methylprop-2-enoate
Molecular FormulaC6H10O3
InChI KeyWOBHKFSMXKNTIM-UHFFFAOYSA-N
SMILESCC(=C)C(=O)OCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Acid value=<2 mg KOH/g
Appearance (Color)Clear colorless to light yellow
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
Stabilizer180 to 220 ppm MEHQ
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only