5-Hydroxyquinoline, 97%
5-Hydroxyquinoline, 97%
5-Hydroxyquinoline, 97%
Thermo Scientific Chemicals

5-Hydroxyquinoline, 97%

CAS: 578-67-6 | C9H7NO | 145.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 156430010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS578-67-6
IUPAC Namequinolin-1-ium-5-olate
Molecular FormulaC9H7NO
InChI KeyGYESAYHWISMZOK-UHFFFAOYSA-N
SMILES[O-]C1=C2C=CC=[NH+]C2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige to brown
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only