Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
Thermo Scientific™

Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate

Color: Clear; Sterility: Sterile; Lid: Without Lid
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsNo. per PackNo. per Case
1565459610 Microplates180 Microplates
168055961 Microplate50 Microplates
Catalog number 156545
Price (USD)
1,011.65
Online exclusive
1,076.00
Save 64.35
Case of 180
No. of Wells:
96
No. per Pack:
10 Microplates
No. per Case:
180 Microplates
Price (USD)
1,011.65
Online exclusive
1,076.00
Save 64.35
Case of 180

Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.

  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Description96F well plate, Nunclon Delta, clear, without lid, Sterile (Pack of 10, Case of 180)
LidWithout Lid
No. per Case180 Microplates
No. per Pack10 Microplates
Packaging10/Pk., 180/Cs.
Product TypeMicroplate
Quantity180/Cs.
Skirt StyleHigh Flange
Volume (Metric) Well400 μL
Array8 x 12
BarcodeCustom Barcoding Available
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNunclon Delta
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 180

Frequently asked questions (FAQs)

What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .