p-Anisylchlorodiphenylmethane, 97%
p-Anisylchlorodiphenylmethane, 97%
p-Anisylchlorodiphenylmethane, 97%
Thermo Scientific Chemicals

p-Anisylchlorodiphenylmethane, 97%

CAS: 14470-28-1 | C20H17ClO | 308.8 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 156561000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14470-28-1
IUPAC Name1-(chlorodiphenylmethyl)-4-methoxybenzene
Molecular FormulaC20H17ClO
InChI KeyOBOHMJWDFPBPKD-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric96.0 to 104.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only