4-Iodoaniline, 99%, Thermo Scientific Chemicals
4-Iodoaniline, 99%, Thermo Scientific Chemicals
4-Iodoaniline, 99%, Thermo Scientific Chemicals

4-Iodoaniline, 99%, Thermo Scientific Chemicals

CAS: 540-37-4 | C6H6IN | 219.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 156610250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS540-37-4
IUPAC Name4-iodoaniline
Molecular FormulaC6H6IN
InChI KeyVLVCDUSVTXIWGW-UHFFFAOYSA-N
SMILESNC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Beige to grey-brown to dark grey
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Melting point61°C to 64°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only