3-Indolylacetonitrile, 97%
3-Indolylacetonitrile, 97%
3-Indolylacetonitrile, 97%
Thermo Scientific Chemicals

3-Indolylacetonitrile, 97%

CAS: 771-51-7 | C10H8N2 | 156.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 156650250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS771-51-7
IUPAC Name2-(1H-indol-3-yl)acetonitrile
Molecular FormulaC10H8N2
InChI KeyDMCPFOBLJMLSNX-UHFFFAOYSA-N
SMILESN#CCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid or solid
Refractive index1.6080 to 1.6110 (20°C, 589 nm)
Appearance (Color)Clear yellow-brown to brown
HPLC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only