2-Isopropylphenol, 97%
2-Isopropylphenol, 97%
2-Isopropylphenol, 97%
Thermo Scientific Chemicals

2-Isopropylphenol, 97%

CAS: 88-69-7 | C9H12O | 136.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
Packaging:
Glass bottle
Catalog number 156685000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS88-69-7
IUPAC Name2-(propan-2-yl)phenol
Molecular FormulaC9H12O
InChI KeyCRBJBYGJVIBWIY-UHFFFAOYSA-N
SMILESCC(C)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to orange
Appearance (Form)Liquid
Appearance (Color)May darken on storage
Infrared spectrumConforms
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only