Isovaleric acid, 99%
Isovaleric acid, 99%
Isovaleric acid, 99%
Thermo Scientific Chemicals

Isovaleric acid, 99%

CAS: 503-74-2 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 156695000
Price (USD)
81.10
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
81.10
Each
Chemical Identifiers
CAS503-74-2
IUPAC Name3-methylbutanoic acid
Molecular FormulaC5H10O2
InChI KeyGWYFCOCPABKNJV-UHFFFAOYSA-N
SMILESCC(C)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3990 to 1.4070 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only