N-Hydroxysuccinimide, 98+%
N-Hydroxysuccinimide, 98+%
N-Hydroxysuccinimide, 98+%
Thermo Scientific Chemicals

N-Hydroxysuccinimide, 98+%

CAS: 6066-82-6 | C4H5NO3 | 115.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 157272500
Price (USD)
274.65
Online Exclusive
305.00
Save 30.35 (10%)
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
274.65
Online Exclusive
305.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS6066-82-6
IUPAC Name1-hydroxypyrrolidine-2,5-dione
Molecular FormulaC4H5NO3
InChI KeyNQTADLQHYWFPDB-UHFFFAOYSA-N
SMILESON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals
AppearanceMay have a tendency to cake
Infrared spectrumConforms
Titration with NaOH98.0 to 101.0 % (On dried substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only