5-Aminoindan, 97%
5-Aminoindan, 97%
5-Aminoindan, 97%
Thermo Scientific Chemicals

5-Aminoindan, 97%

CAS: 24425-40-9 | C9H11N | 133.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 157350500
Price (USD)
337.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
337.00
Each
Chemical Identifiers
CAS24425-40-9
IUPAC Name2,3-dihydro-1H-inden-5-amine
Molecular FormulaC9H11N
InChI KeyLEWZOBYWGWKNCK-UHFFFAOYSA-N
SMILESNC1=CC=C2CCCC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline mass
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only