Fumaronitrile, 95%
Fumaronitrile, 95%
Fumaronitrile, 95%
Fumaronitrile, 95%
Thermo Scientific Chemicals

Fumaronitrile, 95%

CAS: 764-42-1 | C4H2N2 | 78.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 157400250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS764-42-1
IUPAC Name(2E)-but-2-enedinitrile
Molecular FormulaC4H2N2
InChI KeyKYPOHTVBFVELTG-OWOJBTEDSA-N
SMILESN#C\C=C\C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light brown
Melting point93°C to 100°C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only