4-Picoline, 98%
4-Picoline, 98%
4-Picoline, 98%
Thermo Scientific Chemicals

4-Picoline, 98%

CAS: 108-89-4 | C6H7N | 93.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 157600025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-89-4
IUPAC Name4-methylpyridine
Molecular FormulaC6H7N
InChI KeyFKNQCJSGGFJEIZ-UHFFFAOYSA-N
SMILESCC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive index1.5030 to 1.5070 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only