Pyrimidine, 99%
Pyrimidine, 99%
Pyrimidine, 99%
Thermo Scientific Chemicals

Pyrimidine, 99%

CAS: 289-95-2 | C4H4N2 | 80.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 157700050
Price (USD)
99.65
Online exclusive
111.00
Save 11.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
99.65
Online exclusive
111.00
Save 11.35
Each
Chemical Identifiers
CAS289-95-2
IUPAC Namepyrimidine
Molecular FormulaC4H4N2
InChI KeyCZPWVGJYEJSRLH-UHFFFAOYSA-N
SMILESC1=CN=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to orange
Appearance (Form)Liquid or solid
Refractive index1.5010 to 1.5050 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only