Thymine, 99%
Thymine, 99%
Thymine, 99%
Thermo Scientific Chemicals

Thymine, 99%

CAS: 65-71-4 | C5H6N2O2 | 126.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 157851000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS65-71-4
IUPAC Name5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC5H6N2O2
InChI KeyRWQNBRDOKXIBIV-UHFFFAOYSA-N
SMILESCC1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Powder
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only