1,2,4-Trichlorobenzene, 99%, pure
1,2,4-Trichlorobenzene, 99%, pure
1,2,4-Trichlorobenzene, 99%, pure
Thermo Scientific Chemicals

1,2,4-Trichlorobenzene, 99%, pure

CAS: 120-82-1 | C6H3Cl3 | 181.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 157900010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS120-82-1
IUPAC Name1,2,4-trichlorobenzene
Molecular FormulaC6H3Cl3
InChI KeyPBKONEOXTCPAFI-UHFFFAOYSA-N
SMILESClC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<30 APHA
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5700 to 1.5720 (20°C, 589 nm)
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only