Undecylenic aldehyde, 97%
Undecylenic aldehyde, 97%
Undecylenic aldehyde, 97%
Thermo Scientific Chemicals

Undecylenic aldehyde, 97%

CAS: 112-45-8 | C11H20O | 168.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 157990250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-45-8
IUPAC Nameundec-10-enal
Molecular FormulaC11H20O
InChI KeyOFHHDSQXFXLTKC-UHFFFAOYSA-N
SMILESC=CCCCCCCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4400 to 1.4460 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only