Aniline, 99.8%, pure
Aniline, 99.8%, pure
Aniline, 99.8%, pure
Thermo Scientific Chemicals

Aniline, 99.8%, pure

CAS: 62-53-3 | C6H7N | 93.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 158190050
Price (USD)
33.30
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.30
Each
Chemical Identifiers
CAS62-53-3
IUPAC Nameaniline
Molecular FormulaC6H7N
InChI KeyPAYRUJLWNCNPSJ-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.75 %
Appearance (Color)Clear colorless to yellow to orange to light brown
AppearanceMay darken on storage
Refractive index1.5845 to 1.5875 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only