Crotonaldehyde, 99+%
Crotonaldehyde, 99+%
Crotonaldehyde, 99+%
Thermo Scientific Chemicals

Crotonaldehyde, 99+%

CAS: 4170-30-3 | C4H6O | 70.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
158225000500 mLGlass bottle
Catalog number 158225000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4170-30-3
IUPAC Name(2Z)-but-2-enal
Molecular FormulaC4H6O
InChI KeyMLUCVPSAIODCQM-IHWYPQMZSA-N
SMILESCC=C/C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
Free acid=<0.5 % (as Crotonic acid)
Refractive index1.4355 to 1.4375 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only