2-Methyl-1-propanol, 99+%, Extra Pure
2-Methyl-1-propanol, 99+%, Extra Pure
2-Methyl-1-propanol, 99+%, Extra Pure
Thermo Scientific Chemicals

2-Methyl-1-propanol, 99+%, Extra Pure

2-Methyl-1-propanol, 99+%, C4H10O, CAS Number-78-83-1, 1-hydroxymethylpropane, 1-propanol, 2-methyl, 2-methyl-1-propanol, 2-methylpropyl alcohol, i-butyl alcohol, isobutanol, iso-butyl alcohol, isobutyl alcohol, isobutylalkohol, isopropylcarbinol, 2.5L, 99% min. (GC), 01, 373, 108 deg.C, 100.0, 99.0 | CAS: 78-83-1 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
Catalog number 158280010
Price (USD)
68.65
Special offer
Online exclusive
Ends: 28-Dec-2026
80.60
Save 11.95
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
68.65
Special offer
Online exclusive
Ends: 28-Dec-2026
80.60
Save 11.95
Each
Chemical Identifiers
CAS78-83-1
IUPAC Name2-methylpropan-1-ol
Molecular FormulaC4H10O
InChI KeyZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILESCC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Infrared spectrumConforms
Residue after evaporation=<0.005 %
Refractive index1.3945 to 1.3975 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only