4-Phenylphenol, 97%
4-Phenylphenol, 97%
4-Phenylphenol, 97%
Thermo Scientific Chemicals

4-Phenylphenol, 97%

CAS: 92-69-3 | C12H10O | 170.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 158710010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS92-69-3
IUPAC Name[1,1'-biphenyl]-4-ol
Molecular FormulaC12H10O
InChI KeyYXVFYQXJAXKLAK-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point163°C to 168°C
Appearance (Form)Flakes
Appearance (Color)White to beige
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only