Succinic acid, 99%
Succinic acid, 99%
Succinic acid, 99%
Thermo Scientific Chemicals

Succinic acid, 99%

CAS: 110-15-6 | C4H6O4 | 118.09 g/mol
数量:
250 g
500 g
1 kg
2.5 kg
10 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 158745000
価格(JPY)
-
数量:
500 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS110-15-6
IUPAC Namebutanedioic acid
Molecular FormulaC4H6O4
InChI KeyKDYFGRWQOYBRFD-UHFFFAOYSA-N
SMILESOC(=O)CCC(O)=O
さらに表示
Titration with NaOH>=98.5 %
Fumaric and maleic acid=<1 %
Melting point185°C to 190°C
Appearance (Color)White
Water=<1 % (K.F.)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only