Succinic acid, disodium salt hexahydrate, 97%
Succinic acid, disodium salt hexahydrate, 97%
Succinic acid, disodium salt hexahydrate, 97%
Thermo Scientific Chemicals

Succinic acid, disodium salt hexahydrate, 97%

CAS: 6106-21-4 | C4H16Na2O10 | 270.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Catalog number 158750010
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS6106-21-4
IUPAC Namedisodium butanedioate hexahydrate
Molecular FormulaC4H16Na2O10
InChI KeyZBTUYCUNQBRXOR-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Solubility(10 % in water) Clear
Appearance (Form)Crystalline powder or crystals or granules
Titration with HClO496.0 to 103.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only