Succinic Anhydride, 99%
Succinic Anhydride, 99%
Succinic Anhydride, 99%
Thermo Scientific Chemicals

Succinic Anhydride, 99%

Succinic anhydride, 99%, C4H4O3, CAS Number-108-30-5, 2,5-diketotetrahydrofuran, 2,5-furandione, dihydro, butanedioic anhydride, dihydro-2,5-furandione, dihydrofuran-2,5-dione, rikacid sa, succinic acid anhydride, succinic anhydride, succinyl anhydride, succinyl oxide, 1kg, 17,407, 261.0 deg.C | CAS: 108-30-5 | C4H4O3 | 100.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 158762500
Price (USD)
103.65
Online exclusive
115.00
Save 11.35
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
103.65
Online exclusive
115.00
Save 11.35
Each
Chemical Identifiers
CAS108-30-5
IUPAC Nameoxolane-2,5-dione
Molecular FormulaC4H4O3
InChI KeyRINCXYDBBGOEEQ-UHFFFAOYSA-N
SMILESO=C1CCC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or flakes and/or chips
Appearance (Color)White
Infrared spectrumConforms
Melting point116°C to 120°C
Titration by coupling>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only