m-Xylene, 99%, Extra Dry, AcroSeal™
m-Xylene, 99%, Extra Dry, AcroSeal™
m-Xylene, 99%, Extra Dry, AcroSeal™
m-Xylene, 99%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

m-Xylene, 99%, Extra Dry, AcroSeal™

CAS: 108-38-3 | C8H10 | 106.17 g/mol
製品番号(カタログ番号) 158931000
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS108-38-3
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
さらに表示
Water=<0.005 %
GC>=98.5 %
Residue after evaporation=<0.01 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only