ES Series Lab Refrigerators,158 L, Each - FAQs
ES Series Lab Refrigerators,158 L, Each - FAQs
View additional product information for ES Series Lab Refrigerators - FAQs (158R-AEV-TS)
1 product FAQs found
If I order an ES Series Lab Refrigerator or Freezer, what type of plug will it come with?
The ES Series Lab Refrigerator or Freezer will be delivered with the compatible plug type and adapter based on where the unit is shipped.