Triphenylphosphine Sulfide, 99+%
Triphenylphosphine Sulfide, 99+%
Triphenylphosphine Sulfide, 99+%
Thermo Scientific Chemicals

Triphenylphosphine Sulfide, 99+%

CAS: 3878-45-3 | C18H15PS | 294.35 g/mol
製品番号(カタログ番号)数量パッケージ
15903010010 gGlass bottle
製品番号(カタログ番号) 159030100
価格(JPY)
-
数量:
10 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3878-45-3
IUPAC Nametriphenyl-λ⁵-phosphanethione
Molecular FormulaC18H15PS
InChI KeyVYNGFCUGSYEOOZ-UHFFFAOYSA-N
SMILESS=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)White
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Melting point162°C to 164°C
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only