Diethyl oxalate, 99%
Diethyl oxalate, 99%
Diethyl oxalate, 99%
Thermo Scientific Chemicals

Diethyl oxalate, 99%

CAS: 95-92-1 | C6H10O4 | 146.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 159180010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS95-92-1
IUPAC Namediethyl oxalate
Molecular FormulaC6H10O4
InChI KeyWYACBZDAHNBPPB-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Density(20°C) 1.078 to 1.082 kg/l
Color scale=<15 APHA
GC>=98.5 %
Refractive index1.4085 to 1.4105 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only