Product Image
Thermo Scientific™
DRI™ MultiDrug Urine Drugs of Abuse Calibrators
Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ MultiDrug Urine Drugs of Abuse Calibrators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityType
15941 x 10 mLCalibrator 3
15881 x 10 mLCalibrator 1
15891 x 25 mLCalibrator 1
15911 x 10 mLCalibrator 2
15921 x 25 mLCalibrator 2
15951 x 25 mLCalibrator 3
15971 x 10 mLCalibrator 4
15981 x 25 mLCalibrator 4
Catalog number 1594
Price (USD)
-
Quantity:
1 x 10 mL
Type:
Calibrator 3
Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ MultiDrug Urine Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ConcentrationMultianalyte
Packaging1 Bottle
Quantity1 x 10 mL
TypeCalibrator 3
Detectable AnalytesMultiple
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C