5-Aminoisoquinoline, 99%
5-Aminoisoquinoline, 99%
5-Aminoisoquinoline, 99%
Thermo Scientific Chemicals

5-Aminoisoquinoline, 99%

CAS: 1125-60-6 | C9H8N2 | 144.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 159420050
Price (EUR)
95,50
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
95,50
Each
Chemical Identifiers
CAS1125-60-6
IUPAC Nameisoquinolin-5-amine
Molecular FormulaC9H8N2
InChI KeyDTVYNUOOZIKEEX-UHFFFAOYSA-N
SMILESNC1=C2C=CN=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point126°C to 130°C
GC>=98.5 %
Appearance (Color)Beige to yellow-brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only