4,4-Dimethyl-2-pentanone, 99%
4,4-Dimethyl-2-pentanone, 99%
4,4-Dimethyl-2-pentanone, 99%
Acros Organics

4,4-Dimethyl-2-pentanone, 99%

CAS: 590-50-1 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 159710250
also known as 15971
HSN 29141990
Price (INR)
111,270.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
111,270.00
Each
Chemical Identifiers
CAS590-50-1
IUPAC Name4,4-dimethylpentan-2-one
Molecular FormulaC7H14O
InChI KeyAZASWMGVGQEVCS-UHFFFAOYSA-N
SMILESCC(=O)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4025 to 1.4045 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only