2-méthyl-2-oxazoline, 99 %, Thermo Scientific Chemicals
2-méthyl-2-oxazoline, 99 %, Thermo Scientific Chemicals
2-méthyl-2-oxazoline, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-méthyl-2-oxazoline, 99 %, Thermo Scientific Chemicals

CAS: 1120-64-5 | C4H7NO | 85.11 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
100 g
500 g
Conditionnement:
Flacon en verre
Référence 159975000
Prix (EUR)
262,65
Offre exceptionnelle en ligne
292,00
Économisez 29,35 (10%)
Each
Quantité:
500 g
Conditionnement:
Flacon en verre
Grand volume ou format personnalisé
Prix (EUR)
262,65
Offre exceptionnelle en ligne
292,00
Économisez 29,35 (10%)
Each
Identifiants chimiques
CAS1120-64-5
IUPAC Name2-methyl-4,5-dihydro-1,3-oxazole
Molecular FormulaC4H7NO
InChI KeyGUXJXWKCUUWCLX-UHFFFAOYSA-N
SMILESCC1=NCCO1
Voir plus
GC>=98.5 %
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Water=<0.2 % (K.F.)
Infrared spectrumConforms
Voir plus
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only