Rat IgG2b kappa Isotype Control (eB149/10H5), Functional Grade, eBioscience™ - FAQs
Rat IgG2b kappa Isotype Control (eB149/10H5), Functional Grade, eBioscience™ - FAQs
View additional product information for Rat IgG2b kappa Isotype Control (eB149/10H5), Functional Grade, eBioscience™ - FAQs (16-4031-81, 16-4031-050, 16-4031-38, 16-4031-025, 16-4031-85)
2 product FAQs found
Is an isotype control a must for flow cytometry analysis?
No. Many users are using unstained cells in combination with FMO controls to identify their positive populations.
What if I can't find the right isotype control for flow cytomtery?
The flow cytometry field is moving away from using isotype controls as they are not necessarily the most appropriate way to control for non-specific binding. Instead, they are using unstained cells to define the negative population, single stained controls to set compensation, and flow minus one (FMO) controls to set regions and gates.
You may want to consider whether using an isotype control is something you need to do. Here are some references you might want to look at:
-O'Gorman MR, Thomas J. (1999) Isotype controls-time to let go? Cytometry 38:78-80.
-Keeney M, Gratama JW, Chin-Yee IH et al. (1998) Isotype controls in the analysis of lymphocytes and CD34+ stem/progenitor cells by flow cytometry- time to let go! Cytometry 34:280-283.
-Hulspas R, O'Gorman MR, Wood BL et al. (2009) Considerations for the control of background fluorescence in clinical flow cytometry. Cytometry B Clin Cytom 76:355–364.
-Enumeration of Immunologically Defined Cell Populations by Flow Cytometry; Approved Guideline - Second Edition. Clinical and Laboratory Standards Institute, (CLSI). Document H42-A2 Volume 27 No.16, 2007.
-Clinical Flow Cytometric Analysis of Neoplastic Hematolymphoid Cells; Approved Guideline - Second Edition. Clinical and Laboratory Standards Institute, (CLSI). Document H43-A2 Volume 27 No. 11, 2007.
If you do wish to use isotype controls, we offer a wide variety of species and fluorophores, search our Web site under "isotype antibody control."