Product Image
Applied Biosystems™

Ferrule, 1/16

This is a spare part for DNA Sequencers.
Have Questions?
Catalog NumberQuantity
1601 Unit
Catalog number 160
Price (USD)
30.58
Each
Quantity:
1 Unit
Price (USD)
30.58
Each

This is a spare part for DNA Sequencers.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)310 Genetic Analyzer, 3100 Genetic Analyzer
Product TypeFerrule
Quantity1 Unit
Shipping ConditionRoom Temperature
Starting MaterialDNA
Unit SizeEach
Contents & Storage
Store at room temperature.