Nalgene™ Narrow-Mouth Bottles Made of Teflon™ FEP with Closure, 250 mL - FAQs
Nalgene™ Narrow-Mouth Bottles Made of Teflon™ FEP with Closure, 250 mL - FAQs
View additional product information for Nalgene™ Narrow-Mouth Bottles Made of Teflon™ FEP with Closure - FAQs (1600-0016, 1600-0004, 1600-0016PK, 1600-0064TS, 1600-0008PK, 1600-0002, 1600-0002PK, 1600-0064TSPK, 1600-0001PK, 1600-0004PK, 1600-0032, 1600-0008)
1 product FAQs found
Why do you recommend Teflon-coated bottles to store high-value contents such as, active pharmaceutical ingredients (API's), bulk intermediates, and critical reagents?
Teflon is ideal for storing the highest-value contents because it is resistant to most chemicals, temperatures, and processes, allowing for rigorous cleaning procedures such as those that require boiling nitric acid. Teflon also allows you to store contents down to cryogenic temperatures for products requiring ultra-low temperature (below -100 degrees C) storage.
We offer Nalgene Teflon bottles that provide excellent chemical and temperature resistance in a leak-proof design. They are manufactured using quality resins, with the added value of high-level support, making Nalgene Teflon bottles an ideal choice for your high-value contents.