2-Bromoethanesulfonic acid, sodium salt, 98%
2-Bromoethanesulfonic acid, sodium salt, 98%
2-Bromoethanesulfonic acid, sodium salt, 98%
Thermo Scientific Chemicals

2-Bromoethanesulfonic acid, sodium salt, 98%

CAS: 4263-52-9 | C2H4BrNaO3S | 211.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 160030250
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.00
Save 5.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.00
Save 5.35
Each
Chemical Identifiers
CAS4263-52-9
IUPAC Namesodium 2-bromoethane-1-sulfonate
Molecular FormulaC2H4BrNaO3S
InChI KeyHNFOAHXBHLWKNF-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric97.5 to 102.5 % (ex Br)
Titration Argentometric97.5 to 102.5 % (ex Br)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only