Horse Serum, New Zealand origin, 1000 mL - FAQs
Horse Serum, New Zealand origin, 1000 mL - FAQs
View additional product information for Horse Serum, New Zealand origin - FAQs (16050114, 16050130, 16050122)
2 product FAQs found
Does horse serum contain tetracycline?
No. Our horse serum is collected from a controlled herd. Should one of the animals require treatment with an antibiotic, that animal is removed from the drawing program for the full witholding period of any drug administered.