1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%
1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%
Thermo Scientific Chemicals

1,8-Diazabicyclo[5.4.0]undec-7-ene, 98+%

CAS: 6674-22-2 | C9H16N2 | 152.24 g/mol
数量:
100 g
500 g
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 160611000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6674-22-2
IUPAC Name2H,3H,4H,6H,7H,8H,9H,10H-pyrimido[1,2-a]azepine
Molecular FormulaC9H16N2
InChI KeyGQHTUMJGOHRCHB-UHFFFAOYSA-N
SMILESC1CN=C2CCCCCN2C1
さらに表示
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.0 %
Water=<0.5 % (K.F.)
Refractive index1.5210 to 1.5230 (20°C, 589 nm)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only