trans-Methyl crotonate, 96%
trans-Methyl crotonate, 96%
trans-Methyl crotonate, 96%
Thermo Scientific Chemicals

trans-Methyl crotonate, 96%

CAS: 623-43-8 | C5H8O2 | 100.12 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 160841000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS623-43-8
IUPAC Namemethyl (2E)-but-2-enoate
Molecular FormulaC5H8O2
InChI KeyMCVVUJPXSBQTRZ-ONEGZZNKSA-N
SMILESCOC(=O)\C=C\C
さらに表示
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4230 to 1.4270 (20°C, 589 nm)
GC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only