Benzoyl bromide, 97%
Benzoyl bromide, 97%
Benzoyl bromide, 97%
Thermo Scientific Chemicals

Benzoyl bromide, 97%

CAS: 618-32-6 | C7H5BrO | 185.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 160950050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS618-32-6
IUPAC Namebenzoyl bromide
Molecular FormulaC7H5BrO
InChI KeyAQIHMSVIAGNIDM-UHFFFAOYSA-N
SMILESBrC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to light orange
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5880 to 1.5940 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only