4-Nitrobenzyl chloride, 99%
4-Nitrobenzyl chloride, 99%
4-Nitrobenzyl chloride, 99%
Thermo Scientific Chemicals

4-Nitrobenzyl chloride, 99%

CAS: 100-14-1 | C7H6ClNO2 | 171.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
250 g
Packaging:
Glass bottle
Catalog number 161090100
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.50
Save 4.85
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 28-Dec-2026
33.50
Save 4.85
Each
Chemical Identifiers
CAS100-14-1
IUPAC Name1-(chloromethyl)-4-nitrobenzene
Molecular FormulaC7H6ClNO2
InChI KeyKGCNHWXDPDPSBV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(CCl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or needles
Melting point71°C to 74°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only