Rabbit Serum, 100 mL - FAQs
Rabbit Serum, 100 mL - FAQs
View additional product information for Rabbit Serum - FAQs (16120107, 16120099)
2 product FAQs found
What species of rabbit does the Rabbit Serum (Cat. No. 16120099) come from?
The rabbit species is Oryctolagus cuniculus.