trans-1,2-Cyclohexanediol, 97%
trans-1,2-Cyclohexanediol, 97%
trans-1,2-Cyclohexanediol, 97%
Thermo Scientific Chemicals

trans-1,2-Cyclohexanediol, 97%

CAS: 1460-57-7 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 161650100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1460-57-7
IUPAC Name(1R,2R)-cyclohexane-1,2-diol
Molecular FormulaC6H12O2
InChI KeyPFURGBBHAOXLIO-PHDIDXHHSA-N
SMILESO[C@@H]1CCCC[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point100°C to 104°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only