Bovine Serum, New Zealand origin, 1000 mL - FAQs
Bovine Serum, New Zealand origin, 1000 mL - FAQs
View additional product information for Bovine Serum, New Zealand origin - FAQs (16170060, 16170078, 16170086)